Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTG1 Antibody (OTI7H6), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270286APC
Description
BTG1 Monoclonal antibody specifically detects BTG1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
BTG1 | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
OTI7H6 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
B-cell translocation gene 1 protein, B-cell translocation gene 1, proliferative, protein BTG1 | |
Full length human recombinant protein of human BTG1 (NP_001722) produced in E.coli. | |
0.1 mL | |
Angiogenesis, Apoptosis, Cardiovascular Biology, Cell Biology, Cell Cycle and Replication | |
694 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction