Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTG1 Antibody (OTI7H6), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270286UV
Description
BTG1 Monoclonal specifically detects BTG1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
BTG1 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
BTG1 | |
Full length recombinant protein of human BTG1 (NP_001722) produced in E.coli. | |
0.1 mL | |
694.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI7H6 | |
Immunohistochemistry | |
B-cell translocation gene 1 protein, B-cell translocation gene 1, proliferative, protein BTG1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction