Learn More
Invitrogen™ BTG1 Polyclonal Antibody

Description
This antibody is predicted to react with bovine, chicken and rat based on sequence homology.
Specifications
Specifications
| Antigen | BTG1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | BTG1 |
| Gene Accession No. | P62324, P62325 |
| Gene Alias | AI426953; anti-proliferative factor; APRO2; AW546738; B cell translocation gene 1, anti-proliferative; B-cell translocation gene 1 protein; B-cell translocation gene 1, anti-proliferative; BTG anti-proliferation factor 1; Btg1; Protein BTG1 |
| Gene Symbols | BTG1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.