Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | BTG1 |
---|---|
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
BTG1 Polyclonal specifically detects BTG1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
BTG1 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
694 | |
IgG | |
Affinity purified |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
B-cell translocation gene 1 protein, B-cell translocation gene 1, proliferative, protein BTG1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 22-171 of human BTG1 (NP_001722.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title