Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTG2 Antibody (OTI4C10), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271339F
Description
BTG2 Monoclonal specifically detects BTG2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
BTG2 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
BTG2 | |
Full length recombinant protein of human BTG2 (NP_006754) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Immunohistochemistry | |
OTI4C10 | |
Immunohistochemistry | |
B-cell translocation gene 2, BTG family member 2, BTG family, member 2, MGC126063, MGC126064, NGF-inducible proliferative protein PC3, PC3nerve growth factor-inducible proliferative, pheochromacytoma cell-3, protein BTG2, TIS21 | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication | |
7832.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction