Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BTG2 Monoclonal Antibody (OTI1G3)
GREENER_CHOICE

Catalog No. PIMA526944
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526944 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526944 Supplier Invitrogen™ Supplier No. MA526944
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BTG2 Monoclonal Antibody for Western Blot, IHC (P)

The protein encoded by this gene is a member of the BTG/Tob family. This family has structurally related proteins that appear to have antiproliferative properties. This encoded protein is involved in the regulation of the G1/S transition of the cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTG2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1G3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BTG2
Gene Accession No. P78543
Gene Alias AA959598; Agl; An; an-1; APRO1; B cell translocation gene 2, anti-proliferative; B-cell translocation gene 2; B-cell translocation gene 2, anti-proliferative; BTG anti-proliferation factor 2; BTG family member 2; BTG family, member 2; BTG2; cell surface alloantigen; Early induced gene B-cell translocation gene 2; nerve growth factor-inducible anti-proliferative; NGF-inducible anti-proliferative protein PC3; NGF-inducible anti-proliferative putative secreted protein; NGF-inducible protein TIS21; Pc3; pheochromacytoma cell-3; protein BTG2; Tis21
Gene Symbols BTG2
Host Species Mouse
Immunogen Full length human recombinant protein of BTG2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7832
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.