Learn More
BTG2 Mouse anti-Human, Clone: OTI2G5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808151

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a member of the BTG/Tob family. This family has structurally related proteins that appear to have antiproliferative properties. This encoded protein is involved in the regulation of the G1/S transition of the cell cycle.Specifications
BTG2 | |
Monoclonal | |
Unconjugated | |
BTG2 | |
APRO1, PC3, TIS21 | |
Mouse | |
Affinity Chromatography | |
RUO | |
7832 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2G5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P78543 | |
BTG2 | |
Full length human recombit protein of human BTG2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.