Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTG2 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS0031R

Description
The protein encoded by this gene is a member of the BTG/Tob family. This family has structurally related proteins that appear to have antiproliferative properties. This encoded protein is involved in the regulation of the G1/S transition of the cell cycle.Specifications
BTG2 | |
Polyclonal | |
Unconjugated | |
BTG2 | |
AA959598; Agl; An; an-1; APRO1; B cell translocation gene 2, anti-proliferative; B-cell translocation gene 2; B-cell translocation gene 2, anti-proliferative; BTG anti-proliferation factor 2; BTG family member 2; BTG family, member 2; BTG2; cell surface alloantigen; Early induced gene B-cell translocation gene 2; nerve growth factor-inducible anti-proliferative; NGF-inducible anti-proliferative protein PC3; NGF-inducible anti-proliferative putative secreted protein; NGF-inducible protein TIS21; PC3; pheochromacytoma cell-3; Protein BTG2; Tis21 | |
Rabbit | |
Protein A | |
RUO | |
7832 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P78543 | |
BTG2 | |
KLH conjugated synthetic peptide derived from human BTG2 . | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction