Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTG3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $646.00
Specifications
Antigen | BTG3 |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
BTG3 Polyclonal specifically detects BTG3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
BTG3 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
Abundant in neuroepithelium area protein, ANATOB55, B-cell translocation gene 3, B-cell translocation gene10, BTG family member 3, BTG family, member 3, MGC8928, protein BTG3, Protein Tob5, tob55, TOB5TOFA | |
BTG3 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Cell Cycle and Replication | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
10950 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:VDPCEVCCRYGEKNNAFIVASFENKDENKDEISRKVTRALDKVTSDYHSGSSSSDEETSKEMEVKPSSVT | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title