Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BTG3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553788
Description
Immunogen sequence: VDPCEVCCRY GEKNNAFIVA SFENKDENKD EISRKVTRAL DKVTSDYHSG SSSSDEETSK EMEVKPSSVT Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.
Overexpression impairs serum-induced cell cycle progression from the G0/G1 to S phase.
Specifications
BTG3 | |
Polyclonal | |
Unconjugated | |
BTG3 | |
Abundant in neuroepithelium area protein; ANA; APRO4; B cell translocation gene 3; B-cell translocation gene 3; BTG anti-proliferation factor 3; BTG family member 3; BTG family, member 3; Btg3; btg3 {ECO:0000312; Protein BTG3; protein Tob5; rBTG3; RGD:2226}; TOB5; TOB55; TOFA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10950 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14201 | |
BTG3 | |
Recombinant protein corresponding to Human BTG3. Recombinant protein control fragment (Product #RP-91809). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction