Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTG4 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305926G
Description
BTG4 Polyclonal specifically detects BTG4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
BTG4 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
B-cell translocation gene 4, BTG family member 4, MGC33003, PC3BProtein PC3b, protein BTG4 | |
Produced in rabbits immunized with E. coli-derived Human BTG4 fragment. | |
0.1 mL | |
Cell Cycle and Replication | |
54766 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction