Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BTG4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113416
Description
BTG4 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Shows marked antiproliferative activity, being able to induce G(1) arrest.
Specifications
BTG4 | |
Polyclonal | |
Unconjugated | |
Btg4 | |
APRO3; B cell translocation gene 4; B-cell translocation gene 4; BTG anti-proliferation factor 4; BTG family member 4; BTG4; C86116; PC3B; Protein BTG4; protein PC3b; SCIR-27 | |
Rabbit | |
Protein G | |
RUO | |
54766, 56057 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
2.63 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
O70552, Q9NY30 | |
Btg4 | |
Recombinant Human Protein BTG4 protein (1-206AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction