Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTK Antibody (3E5), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | BTK |
---|---|
Clone | 3E5 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
BTK Monoclonal antibody specifically detects BTK in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
BTK | |
ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Agammaglobulinaemia tyrosine kinase, AGMX1MGC126262, AT, ATKIMD1, B-cell progenitor kinase, BPK, Bruton agammaglobulinemia tyrosine kinase, Bruton tyrosine kinase, dominant-negative kinase-deficient Brutons tyrosine kinase, EC 2.7.10, EC 2.7.10.2, PSCTK1, tyrosine-protein kinase BTK, XLAMGC126261 | |
A synthesized peptide derived from Human BTK [UniProt Q06187] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
3E5 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Apoptosis, Immunology, Innate Immunity, Phospho Specific | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
695 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title