Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTK Antibody (7F12H4), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11087047H
Description
BTK Monoclonal specifically detects BTK in Human, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
BTK | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
BTK | |
Purified recombinant C terminal fragment of BTK expressed in E. coli. [UniProt# Q06187] | |
0.1 mL | |
Apoptosis, Immunology, Innate Immunity, Phospho Specific | |
695 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
7F12H4 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Agammaglobulinaemia tyrosine kinase, AGMX1MGC126262, AT, ATKIMD1, B-cell progenitor kinase, BPK, Bruton agammaglobulinemia tyrosine kinase, Bruton tyrosine kinase, dominant-negative kinase-deficient Brutons tyrosine kinase, EC 2.7.10, EC 2.7.10.2, PSCTK1, tyrosine-protein kinase BTK, XLAMGC126261 | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
Primary | |
Human, Primate | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction