Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Btk Monoclonal Antibody (7F12H4)
GREENER_CHOICE

Catalog No. PIMA515337
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515337 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515337 Supplier Invitrogen™ Supplier No. MA515337
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15337 targets BTK in indirect ELISA, IF, IHC, and WB applications and shows reactivity with Human and Non-human primate samples. The MA5-15337 immunogen is purified recombinant fragment of BTK expressed in E. Coli. MA5-15337 detects BTK which has a predicted molecular weight of approximately 77kDa.

Bruton tyrosine kinase (BTK) is a cytoplasmic tyrosine kinase belonging to the SRC-related TEC subfamily of tyrosine kinases. Mutations in the BTK gene have been linked to severe developmental blocks in human B-cell ontogeny and immunodeficiency disorders. It has recently been shown to interact with members of the toll-like receptor (TLR) family such as TLR4, 6, 8, and 9. The TLRs are critical molecules in both the innate and adaptive immunity and can recognize diverse microbial pathogens. BTK has also been shown to interact with key proteins involved in TLR4 signal transduction such as MyD88, TIRAP, and IRAK, but not TRAF-6, suggesting that BTK is involved in lipopolysaccharide signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Btk
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 7F12H4
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene BTK
Gene Accession No. Q06187
Gene Alias agammaglobulinaemia tyrosine kinase; agammaglobulinemia tyrosine kinase; AGMX1; AI528679; AT; ATK; B-cell progenitor kinase; BPK; Bruton agammaglobulinemia tyrosine kinase; Bruton tyrosine kinase; Bruton's tyrosine kinase; BTK; dominant-negative kinase-deficient Brutons tyrosine kinase; IMD1; Kinase EMB; MGC126261; MGC126262; OTTHUMP00000023677; PSCTK1; RP1-164F3.2; truncated Bruton agammaglobulinemia tyrosine kinase; tyrosine-protein kinase BTK; Tyrosine-protein kinase BTK (Brutons tyrosine kinase) (Agammaglobulinaemia tyrosine kinase) (ATK) (B cell progenitor kinase) (BPK) (Kinase EMB); tyrosine-protein kinase BTK isoform (lacking exon 13 to 17); tyrosine-protein kinase BTK isoform (lacking exon 14); xid; XLA; X-linked immune deficiency
Gene Symbols BTK
Host Species Mouse
Immunogen Purified recombinant fragment of BTK expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 695
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.