Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTK, p Tyr223 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178295MFV450
Description
BTK Polyclonal antibody specifically detects BTK in Human, Mouse samples. It is validated for Flow CytometrySpecifications
BTK | |
Polyclonal | |
Flow Cytometry | |
Agammaglobulinaemia tyrosine kinase, AGMX1MGC126262, AT, ATKIMD1, B-cell progenitor kinase, BPK, Bruton agammaglobulinemia tyrosine kinase, Bruton tyrosine kinase, dominant-negative kinase-deficient Brutons tyrosine kinase, EC 2.7.10, EC 2.7.10.2, PSCTK1, tyrosine-protein kinase BTK, XLAMGC126261 | |
A synthetic phosphorylated peptide made to a peptide surrounding amino acid 223 of the human BTK protein [UniProt Q06187] | |
0.1 mL | |
Apoptosis, Immunology, Innate Immunity, Phospho Specific | |
695 | |
Store at 4C in the dark. | |
IgG |
Flow Cytometry | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction