Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTK, p Tyr223 Antibody (SR2206), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | BTK |
---|---|
Clone | SR2206 |
Dilution | Western Blot 1:1000-1:2000, Immunoprecipitation 1:20 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
BTK Monoclonal antibody specifically detects BTK in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
BTK | |
Western Blot 1:1000-1:2000, Immunoprecipitation 1:20 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Agammaglobulinaemia tyrosine kinase, AGMX1MGC126262, AT, ATKIMD1, B-cell progenitor kinase, BPK, Bruton agammaglobulinemia tyrosine kinase, Bruton tyrosine kinase, dominant-negative kinase-deficient Brutons tyrosine kinase, EC 2.7.10, EC 2.7.10.2, PSCTK1, tyrosine-protein kinase BTK, XLAMGC126261 | |
A synthesized peptide derived from human phospho-BTK [Tyr223] (Uniprot #: Q06187) | |
Primary | |
Store at -20 degrees C. Avoid freeze/thaw cycles. |
SR2206 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Apoptosis, Immunology, Innate Immunity, Phospho Specific | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
695 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title