Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTK Rabbit anti-Human, Mouse, APC, Clone: B4, Abnova™

Numéro de catalogue. 89335656 missing translation for 'brandsLinkText'
Modifier l'affichage
Click to view available options
Quantity:
10 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
89-335-656 10 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 89-335-656 Fournisseur Abnova Corporation Code fournisseur MAB18822
Ajouter au panier
Ajouter au panier

Rabbit Monoclonal Antibody.

BTK Monoclonal antibody specifically detects BTK in Human, Mouse samples. It is validated for Flow Cyt.

Spécifications

Antigen BTK
Applications Flow Cytometry
Classification Monoclonal
Clone B4
Conjugate APC
Description Bruton agammaglobulinemia tyrosine kinase
Gene BTK
Gene Alias AGMX1, AT, ATK, BPK, IMD1, MGC126261, MGC126262, PSCTK1, XLA
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding Y223 of human BTK
Quantity 10 Reactions
Primary or Secondary Primary
Gene ID (Entrez) 695
Target Species Human, Mouse
Content And Storage Store at 4°C
Form Liquid
Isotype IgG1 κ
Afficher plus de résultats Afficher moins de résultats
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.