Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTK, Rabbit anti-Human, SureLight 488, Clone: B4, Abnova™
Rabbit Monoclonal Antibody
Supplier: Abnova Corporation MAB18828
Specifications
BTK | |
Monoclonal | |
SureLight 488 | |
In PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide). | |
AGMX1/AT/ATK/BPK/IMD1/MGC126261/MGC126262/PSCTK1/XLA | |
Rabbit | |
10 Reactions | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |
Flow Cytometry | |
B4 | |
Flow Cytometry (5 uL/106 cells) The optimal working dilution should be determined by the end user. | |
Bruton agammaglobulinemia tyrosine kinase | |
BTK | |
A synthetic phosphopeptide corresponding to residues surrounding Y223 of human BTK. | |
RUO | |
695 | |
Store at 4°C. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction