Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTLA/CD272 Antibody (2161A), Allophycocyanin/Cy7, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB33542APCCY7
Description
BTLA/CD272 Monoclonal antibody specifically detects BTLA/CD272 in Human samples. It is validated for Flow CytometrySpecifications
BTLA/CD272 | |
Monoclonal | |
APC-Cyanine7 | |
PBS | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
2161A | |
Flow Cytometry | |
B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743 | |
Mouse myeloma cell line, NS0-derived human BTLA, Ile25-Ser150, Accession # AAP44003 | |
0.1 mL | |
Cell Cycle and Replication, Phospho Specific | |
151888 | |
Store at 4C in the dark. Do not freeze. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction