Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTLA/CD272 Antibody (753131), Alexa Fluor™ 594, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB7600T
Description
BTLA/CD272 Monoclonal antibody specifically detects BTLA/CD272 in Mouse samples. It is validated for Western BlotSpecifications
BTLA/CD272 | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot | |
753131 | |
Western Blot | |
B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743 | |
Mouse myeloma cell line NS0-derived recombinant mouse BTLA, Met1-Pro176, Accession # Q7TSA3 | |
0.1 mL | |
Cell Cycle and Replication | |
151888 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction