Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTLA/CD272 Antibody (BTLA/1528), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308619AF700
Description
BTLA/CD272 Monoclonal antibody specifically detects BTLA/CD272 in Human samples. It is validated for ELISASpecifications
BTLA/CD272 | |
Monoclonal | |
Alexa Fluor 700 | |
B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743 | |
Recombinant extracellular human BTLA/CD272 protein (Uniprot: Q7Z69) | |
100 μg | |
Cell Cycle and Replication | |
151888 | |
Store at 4°C in the dark. | |
IgG1 κ |
ELISA | |
BTLA/1528 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction