Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTLA/CD272 Antibody (HMBT-6B2), DyLight 350, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NBP281117UV
Description
BTLA/CD272 Monoclonal specifically detects BTLA/CD272 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunoblotting.Specifications
BTLA/CD272 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
BTLA | |
This antibody was raised by immunising Armenian hamsters with mouse BTLA/CD272:Fc fusion protein. | |
0.1 mL | |
Cell Cycle and Replication, Phospho Specific | |
151888 | |
Mouse | |
Purified |
Western Blot, Flow Cytometry, Immunoblot | |
HMBT-6B2 | |
Western Blot, Flow Cytometry, Immunoblotting | |
B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743 | |
Armenian Hamster | |
Protein A purified | |
RUO | |
Primary | |
This antibody is specific for B and T lymphocyte attenuator (BTLA), which functions as a negative regulator of T cell activation and proliferation, and attenuates B cell proliferation upon associating with its known ligand, herpes virus entry mediator (HVEM). | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction