Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTLA/CD272 Antibody (MIH26), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314617JF646
This item is not returnable.
View return policy
Description
BTLA/CD272 Monoclonal antibody specifically detects BTLA/CD272 in Human, Non-human Primate samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
BTLA/CD272 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG2a κ |
Flow Cytometry, Immunofluorescence | |
MIH26 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743 | |
Cells transfected with human BTLA/CD272 | |
0.1 mL | |
Cell Cycle and Replication | |
151888 | |
Store at 4°C in the dark. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction