Learn More
BTLA Mouse anti-Human, Clone: OTI4C2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505547

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
BTLA or B and T-lymphocyte attenuator is a member of the co-inhibitory receptors of the CD28 superfamily. BTLA is a lymphocyte inhibitory receptor which inhibits lymphocytes during immunes responses. BTLA is constitutively expressed on most CD4+ and CD8+ T cells and its expression progressively decreases upon T cell activation. It remains expressed on Th1 cells, but not Th2 cells. BTLA is a unique co-receptor that interacts with the tumor necrosis factor receptor superfamily member herpesvirus entry mediator (HVEM). This interation is an important pathway regulating lymphocyte activation and/or homeostasis in the immune response.Specifications
BTLA | |
Monoclonal | |
Unconjugated | |
BTLA | |
BTLA1, CD272 | |
Mouse | |
Affinity Chromatography | |
RUO | |
151888 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4C2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q7Z6A9 | |
BTLA | |
Full length human recombit protein of human BTLA produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.