Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTN1A1/Butyrophilin Antibody (2151C), Alexa Fluor™ 647, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals MAB8467AF647
Description
BTN1A1/Butyrophilin Monoclonal specifically detects BTN1A1/Butyrophilin in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
BTN1A1/Butyrophilin | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
BTN1A1 | |
Mouse myeloma cell line NS0-derived recombinant human BTN1A1/Butyrophilin | |
100 μL | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
2151C | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
BTBTNbK14H9.2 (butyrophilin, subfamily 1, member A1), butyrophilin subfamily 1 member A1, butyrophilin, subfamily 1, member A1 | |
Rabbit | |
Protein A or G purified | |
RUO | |
696 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction