Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTN1A1/Butyrophilin Antibody (OTI7B5), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270299FR
Description
BTN1A1/Butyrophilin Monoclonal specifically detects BTN1A1/Butyrophilin in Human, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
BTN1A1/Butyrophilin | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
BTN1A1 | |
Full length recombinant protein of human BTN1A1/Butyrophilin (NP_001723) produced in HEK293T cells. | |
0.1 mL | |
696.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI7B5 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
BTBTNbK14H9.2 (butyrophilin, subfamily 1, member A1), butyrophilin subfamily 1 member A1, butyrophilin, subfamily 1, member A1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Canine | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction