Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BTN2A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542931
Description
Peptide sequence: SVALVIHNIT AQENGTYRCY FQEGRSYDEA ILHLVVAGLG SKPLISMRGH Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 86%; Horse: 82%; Human: 100%; Mouse: 91%; Pig: 100%; Rabbit: 100%; Rat: 91%.
The specific functin of this protein remains unknown. This gene is a member of the BTN2 subfamily of genes, which encode proteins belonging to the butyrophilin protein family. The gene is located in a cluster on chromosome 6, consisting of seven genes belonging to the expanding B7/butyrophilin-like group, a subset of the immunoglobulin gene superfamily. The encoded protein is an integral plasma membrane B box protein involved in lipid, fatty-acid and sterol metabolism.
Specifications
BTN2A1 | |
Polyclonal | |
Unconjugated | |
BTN2A1 | |
BK14H9.1; BT2.1; BTF1; BTN2.1; BTN2A1; Butyrophilin; Butyrophilin subfamily 2 member A1; butyrophilin, subfamily 2, member A1; DJ3E1.1; FLJ36567 | |
Rabbit | |
Affinity chromatography | |
RUO | |
11120 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot, Surface Plasmon Resonance | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q7KYR7 | |
BTN2A1 | |
Synthetic peptide directed towards the N-terminal of human BTN2A1 (aa 107-156). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction