Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTN2A1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA587883
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587883 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587883 Supplier Thermo Scientific Supplier No. PA587883

Rabbit Polyclonal Antibody

Immunogen sequence: FIVVGPTDPI LATVGENTTL RCHLSPEKNA EDMEVRWFRS QFSPAVFVYK GGRERTEEQM EEYRGRTTFV SKDISRGSVA LVIHNITAQE NGTYRCYFQE GRSYDEAILH LVVAGLGSKP LISMRGHEDG GIRLECISRG WYPKPLTVWR DPYGGVAPAL KEVSMPDADG LFMVTTAVII RDKSVRNMSC SINNTLLGQK KESVIFIPES F; Positive Samples: U-251MG, HT-1080, HT-29, LO2, A-549, 293T; Cellular Location: Membrane, Single-pass type I membrane protein

The specific functin of this protein remains unknown.This gene is a member of the BTN2 subfamily of genes, which encode proteins belonging to the butyrophilin protein family. The gene is located in a cluster on chromosome 6, consisting of seven genes belonging to the expanding B7/butyrophilin-like group, a subset of the immunoglobulin gene superfamily. The encoded protein is an integral plasma membrane B box protein involved in lipid, fatty-acid and sterol metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTN2A1
Applications Western Blot
Classification Polyclonal
Concentration 4.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BTN2A1
Gene Accession No. Q7KYR7
Gene Symbols BTN2A1
Host Species Rabbit
Immunogen Recombit fusion protein containing a sequence corresponding to amino acids 30-240 of human BTN2A1
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11120
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.