Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BTN2A3P Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549178
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549178 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549178 Supplier Invitrogen™ Supplier No. PA549178
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BTN2A3P Polyclonal Antibody for Western Blot

The butyrophilin (BTN) genes are a group of major histocompatibility complex (MHC)-associated genes that encode type I membrane proteins with 2 extracellular immunoglobulin (Ig) domains and an intracellular B30.2 (PRYSPRY) domain. Three subfamilies of human BTN genes are located in the MHC class I region: the single-copy BTN1A1 gene (MIM 601610) and the BTN2 (e.g., BTN2A3) and BTN3 (e.g., BNT3A1; MIM 613593) genes, which have undergone tandem duplication, resulting in 3 copies of each (summary by Smith et al., 2010 [PubMed 20208008]).[supplied by OMIM, Nov 2010].
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTN2A3P
Applications Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene BTN2A3P
Gene Accession No. Q96KV6
Gene Alias BTN2.3; BTN2A3; BTN2A3P; butyrophilin subfamily 2 member A3, pseudogene
Gene Symbols BTN2A3P
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 293-319 amino acids from the Central region of human BTN2A3.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54718
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.