Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTN3A2 Antibody (OTI1A6), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271678PE
Description
BTN3A2 Monoclonal antibody specifically detects BTN3A2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
BTN3A2 | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
11118 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
OTI1A6 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
BT3.2, BT3.3, BTF3, BTF4butyrophilin protein, butyrophilin subfamily 3 member A2, butyrophilin, subfamily 3, member A2, FLJ40011 | |
Full length human recombinant protein of human BTN3A2 (NP_008978) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction