Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BTN3A2 Monoclonal Antibody (OTI1A6)
GREENER_CHOICE

Catalog No. PIMA525004
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525004 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525004 Supplier Invitrogen™ Supplier No. MA525004
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BTN3A2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

BTN3A2 encodes a member of the immunoglobulin superfamily, which resides in the juxta-telomeric region of the major histocompatability class 1 locus and is clustered with the other family members on chromosome 6. The encoded protein may be involved in the adaptive immune response. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTN3A2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI1A6
Concentration 1.00 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BTN3A2
Gene Accession No. P78410
Gene Alias BT3.2; BTF3; BTF4; BTN3.2; BTN3A2; butyrophilin protein; butyrophilin subfamily 3 member A2; butyrophilin, subfamily 3, member A2; CD277
Gene Symbols BTN3A2
Host Species Mouse
Immunogen Full length human recombinant protein of BTN3A2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11118
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.