Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTN3A3 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300127C
Description
BTN3A3 Polyclonal specifically detects BTN3A3 in Human samples. It is validated for ELISA.Specifications
BTN3A3 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
10384 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
DyLight 650 | |
BTF3butyrophilin 3, butyrophilin subfamily 3 member A3, butyrophilin, subfamily 3, member A3 | |
Produced in rabbits immunized with purified, recombinant BTN3A3 (O00478-1; Met1-Trp248). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction