Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BTN3A3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548885
Description
BTN3A3 Polyclonal Antibody for Western Blot
Plays a role in T-cell responses in the adaptive immune response.
Specifications
BTN3A3 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
O00478 | |
BTN3A3 | |
KLH conjugated synthetic peptide between 67-96 amino acids from the N-terminal region of human BTN3A3. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
BTN3A3 | |
BTF3; BTN3.3; BTN3A3; butyrophilin 3; butyrophilin subfamily 3 member A3; butyrophilin subfamily 3 member A3 secreted isoform; butyrophilin, subfamily 3, member A3 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
10384 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction