Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BTNL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598979
Description
BTNL2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
BTNL2 (butyrophilin-like protein 2), also known as BTL-II, is a 455 amino acid single-pass type II membrane protein that acts as a negative regulator of T cell proliferation. Expressed in thymus, brain, kidney, heart, liver, small intestine, testis, ovary, pancreas and leukocytes, BTNL2 exists as at least six alternatively spliced isoforms, which are encoded by a gene located on human chromosome 6. Expression of BTNL2 isoform 3 is associated with susceptibility to sarcoidosis type 2 (SS2), an immune disorder that causes chronic inflammatory granulomatous lesions.
Specifications
BTNL2 | |
Polyclonal | |
Unconjugated | |
BTNL2 | |
Btl2; BTL-II; BTLN2; BTN7; BTNL2; butyrophilin like 2; butyrophilin-like 2; butyrophilin-like 2 (MHC class II associated); butyrophilin-like protein 2; butyrophylin-like MHC class II associated; Gm315; HSBLMHC1; NG9; SS2 | |
Rabbit | |
Protein G | |
RUO | |
547431, 56244 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
4 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
O70355, Q9UIR0 | |
BTNL2 | |
Recombinant Human Butyrophilin-like protein 2 protein (26-202AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction