Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BTNL8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119886
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119886 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119886 Supplier Invitrogen™ Supplier No. PA5119886
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: Rat brain tissue lysate, mouse testis tissue lysate, human stomach tissue. Subcellular Location: Membrane.

BTNL8 is a single-pass type I membrane protein. It belongs to the immunoglobulin superfamily, BTN/MOG family. BTNL8 contains 1 B30.2/SPRY domain and 1 Ig-like V-type domain. The exact function of BTNL8 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTNL8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene BTNL8
Gene Accession No. Q6UX41
Gene Alias B7-H5 costimulatory molecule; BTN9.2; BTNL8; butyrophilin like 8; butyrophilin-like 8; butyrophilin-like protein 8; UNQ702/PRO1347
Gene Symbols BTNL8
Host Species Rabbit
Immunogen Synthetic peptide within human BTNL8 aa 480-500.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 406160, 79908
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.