Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BTNL9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543947
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543947 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543947 Supplier Invitrogen™ Supplier No. PA543947
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: EIRWFRSQTF NVVHLYQEQQ ELPGRQMPAF RNRTKLVKDD IAYGSVVLQL Sequence homology: Horse: 79%; Human: 100%.

The specific function of BTNL9 is not yet known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTNL9
Applications Western Blot, Surface Plasmon Resonance
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene BTNL9
Gene Accession No. Q6UXG8
Gene Alias BTN3; BTN8; BTNL9; butyrophilin 3; butyrophilin like 9; butyrophilin-like 9; butyrophilin-like protein 9; UNQ1900/PRO4346; VDLS1900
Gene Symbols BTNL9
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human BTNL9 (aa 71-120).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153579
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.