Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BTRC Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144646
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144646 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144646 Supplier Invitrogen™ Supplier No. PA5144646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: HepG2 cell lysate, Hela cell lysate, 293T cell lysate, mouse testis tissue lysate, rat brain tissue lysate, mouse brain tissue lysate, A549, human colon tissue, mouse large intestine tissue. Predicted band size: 69 kDa Subcellular Location: Cytoplasm, Nucleus.

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbws class; in addition to an F-box, this protein contains multiple WD-40 repeats. This protein is homologous to Xenopus bTrCP1, yeast Met30, Neurospora Scon2 and Drosophila Slimb proteins. It interacts with HIV-1 Vpu and connects CD4 to the proteolytic machinery. It also associates specifically with phosphorylated IkappaBalpha and beta-catenin destruction motifs, probably functioning in multiple transcriptional programs by activating the NF-kappaB pathway and inhibiting the beta-catenin pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTRC
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene BTRC
Gene Accession No. Q3ULA2, Q9Y297
Gene Alias beta-transducin repeat containing E3 ubiquitin protein ligase; beta-transducin repeat containing protein; Beta-transducin repeat-containing protein; betaTrCP; Beta-TrCP; Beta-TrCP protein E3RS-IkappaB; Beta-Trcp1; Btrc; BTRCP; b-TrCP; bTrCP1; E3RSIkappaB; E3RS-IkappaB; epididymis tissue protein Li 2a; F-box and WD repeats protein beta-TrCP; F-box and WD-repeat protein 1B; F-box/WD repeat-containing protein 1A; Fbw1; FBW1A; FBXW1; FBXW1A; FWD1; HOS; Kiaa4123; LOW QUALITY PROTEIN: F-box/WD repeat-containing protein 1A; mE3RS-IkappaB; pIkappaBalpha-E3 receptor subunit; pIkappaB-E3 receptor subunit; SCF b-TRCP; Slimb; Ubiquitin ligase FWD1
Gene Symbols BTRC
Host Species Rabbit
Immunogen Recombinant protein within Human Beta TRCP aa 1-200/605.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12234, 361765, 8945
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.