Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BTT 3033
GSA_VAGREENER_CHOICE

Catalog No. 472410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-241-0 10 mg
47-245-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 47-241-0 Supplier Tocris Bioscience™ Supplier No. 4724/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective inhibitor of integrin α2β1

BTT 3033 is a selective inhibitor of integrin α2β1 (EC50 = 130 nM for α2β1 binding to collagen I). Binds to the α2I domain. Exhibits selectivity for α2β1 over integrins α3β1, α4β1, α5β1 and αv. BTT 3033 inhibits platelet aggregation to collagen I coated capillaries under flow and inhibits binding of α2-expressing CHO cells to collagen I under shear stress conditions. Neurogenic and thromboxane A2-induced human prostate smooth muscle contraction are also inhibited by BTT 3033.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Quantity 10 mg
Product Type BTT 3033
Purity >98%
Molecular Weight (g/mol) 465.5
Inhibitors BTT 3033

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.