Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ btuB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144445
Description
btuB Polyclonal Antibody for Western Blot, ELISA
Involved in the active translocation of vitamin B12 (cyanocobalamin) across the outer membrane to the periplasmic space. It derives its energy for transport by interacting with the trans-periplasmic membrane protein TonB.
Specifications
btuB | |
Polyclonal | |
Unconjugated | |
btuB; Vitamin B12 transporter BtuB | |
Recombinant Salmonella enterica (Salmonella choleraesuis) Vitamin B12 transporter BtuB (31-120aa). | |
100 μL | |
Primary | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
1.72 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Rabbit | |
Protein G | |
RUO | |
Bacteria | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction