Learn More
BTX™ Tweezertrodes™ Electrodes Accessory: Cable
Catalog No. BTX524 Shop All BTX Harvard Products
Change view
Click to view available options
Connection Type:
Banana Cables
Catalog No. Connection Type
BTX524 Banana Cables
1 options
Catalog No. BTX524 Supplier BTX™ Supplier No. 450204
Add to Cart
Add to Cart
Cable for use with Tweezertrodes Electrodes.
  • For connection to electroporation generators
  • 1.1 m in length
Specifications
For Use With (Application) In-Vivo Drug or Gene Delivery, Gene Therapy, Protein Delivery
For Use With (Equipment) BTX Tweezertrodes, L-Shaped Genetrode Electrodes, Adherent Cell Electrode
Description Adapter Banana Plug Cables, Red and Black (for use with Tweezertrodes and Adherent Cell Electrode)
Includes Red and Black Cable Set
Length (English) Cable 3.5 ft.
Length (Metric) Cable 1.1 m
Sterility Non-sterile
Connection Type Banana Cables
Pulse Form Exponential Decay Wave, Square Wave