Learn More
BTX™ ECM™ 2001 Electroporation Electro Cell Fusion and Electroporation System
Shop All BTX Harvard Products
Click to view available options
:
Electro Cell Fusion System; Includes: Generator, Micro-Slides, Meander Fusion Chamber, Flat Electrode/Divergent Field, Electrode Adapter, Connection Cable, Safety Stand, Cuvettes, Cuvette Rack
Description
Applications
- Cell Fusion
- Nuclear Transfer
- Embryo Manipulation
- Hybridoma Production
- Mammalian Cell Transfection
- Plant Tissue Transfection
- Plant Protoplast Fusion
Includes:
- ECM 2001 Generator, Micro-Slides 450, 453, Meander Fusion Chamber 454, Flat Electrode/Divergent Field 484, Electrode Adapter, Connection Cable, Safety Stand 630B, Cuvettes 1 mm, 2 mm, 4 mm, pkg. of 30 (10 each), Cuvette Rack 660
Specifications
Specifications
| For Use With (Application) | Animal Embryo Cloning, Hybridoma and Quadroma Production, Plant Protoplast Fusion for Transgenic Plant Generations and Electroporation of Plant Cells, Mammalian Cells, Bacteria, Yeast and Fungi |
| Includes | Generator, Micro-Slides, Meander Fusion Chamber, Flat Electrode/Divergent Field, Electrode Adapter, Connection Cable, Safety Stand, Cuvettes, Cuvette Rack |
| Quantity | 30 Pk. |
| Voltage | 100/115 V |