Learn More
BTX™ BTXPress™ Electroporation Buffer
Shop All BTX Harvard ProductsDescription
BTXPress™ Electroporation Solution is the first electroporation reagent that meets all of your high performance transfection needs without sacrificing control over your experiment or your budget. It offers increased numbers of transfections per kit compared to other commercial alternatives, providing higher value to you. As a universal solution, the BTXPress™ Electroporation Solution can be used in other electroporators, including the Amaxa™, to deliver similar results without the typical high cost associated with these buffer kits. The BTXPress™ Electroporation Solution is offered as a kit including the BTX™ cuvettes Plus with transfer pipettes or as a buffer alone.
- Efficient electroporation transfection
- High efficiency transfection of difficult cell lines
- High cell viability
- Low toxicity resulting in improved cell viability
- Single buffer for all cell types
- One buffer used in place of standard electroporation buffers for all mammalian cell types
Specifications
Specifications
| Buffer | Universal Electroporation Buffer |
| Content And Storage | 2°C to 8°C |
| Color | Clear |
| Description | BTXpress Solution, 2.0 ml Bottle for up to 20 Reactions |
| Cell Type | Mammalian |
| Format | Bottle |
| Sample Type | Cells |
| Promoter | None (Promoterless) |
| Transfection Technique | Electroporation |
| For Use With (Equipment) | BTX™ and other electroporators |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.