Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bu-1 Monoclonal Antibody (AV20), FITC
GREENER_CHOICE

Catalog No. PIMA528701
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528701 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528701 Supplier Invitrogen™ Supplier No. MA528701
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bu-1 Monoclonal Antibody for Flow

Bu-1, a product of the Bu-1a and Bu-1b alleles, is a type I transmembrane glycosylated disulfide-linked homodimer. The monoclonal antibody AV20 recognizes a monomorphic determinant on the Bu-1 B cell-associated alloantigens of both RPL 6(3) (Bu-1a) and 7(2) (Bu-1b) lines of inbred chickens. The antibody also identifies B cells in partially inbred birds. Bu-1 is found on 85-90% of bursal cells, 2-8% of thymocytes, 15-27% of spleen cells, and 2-18% of peripheral blood cells. It is also expressed on subsets of macrophages and monocytes but not on granulocytes, erythrocytes, or thrombocytes. The monoclonal antibody AV20 does not cross react with turkey cells when analyzed by flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bu-1
Applications Flow Cytometry
Classification Monoclonal
Clone AV20
Concentration 0.5 mg/mL
Conjugate FITC
Formulation PBS with <0.1% sodium azide; pH 7.4
Gene LOC396098
Gene Accession No. 0
Gene Alias B6.1; B6.2; Bu-1; Bu-1b; ChB6; LOC396098
Gene Symbols LOC396098
Host Species Mouse
Immunogen Chicken peripheral blood lymphocytes and splenocytes.
Purification Method Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 396098
Target Species Chicken
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.