Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Bu-1b Monoclonal Antibody (5-11G2)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA528759
Description
Bu-1b Monoclonal Antibody for Flow
Bu-1b, a product of the Bu-1b allele, is expressed on the cell surface as a disulfide-linked homodimer. In 10-week-old H.B15 and H.B2 chickens, Bu-1 is found on 85-90% of bursal cells, 2-8% of thymocytes, 15-27% of spleen cells, and 2-18% of peripheral blood cells. It is also expressed on subsets of macrophages and monocytes but not on granulocytes, erythrocytes, or thrombocytes. In chickens heterozygous for the Bu-1 alleles (Bu-1a/b), Bu-1a does not exhibit allelic exclusion. The monoclonal antibody 5-11G2 reacts with lymphoid cells from CHA and H.B14Ab strains but does not react with cells from H.B14Aa.
Specifications
Bu-1b | |
Monoclonal | |
0.5 mg/mL | |
BBS with proprietary stabilizer and no preservative; pH 8.2 | |
0 | |
LOC396098 | |
Bursa cells from two- to five-week-old CHA (Bu-1b) strain chickens. | |
1 mL | |
Primary | |
Chicken | |
Antibody | |
IgG1 κ |
Flow Cytometry | |
5-11G2 | |
Unconjugated | |
LOC396098 | |
B6.1; B6.2; Bu-1; Bu-1b; ChB6.2; LOC396098 | |
Mouse | |
purified | |
RUO | |
396098 | |
4°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction