Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ BUB1/BUB3 (Human) Recombinant Protein
Shop All Abnova Corporation Products
Click to view available options
Quantity:
5 μg
Description
Human BUB1/BUB3 (NP_004327/NP_004716) full length recombinant protein with GST/DYKDDDDK-tag at N-terminal using baculovirus expression system.
Specifications
Specifications
For Use With (Application) | Functional Study |
Gene ID (Entrez) | 699/9184 |
Name | budding uninhibited by benzimidazoles 1 homolog (yeast) |
Quality Control Testing | The purity was assessed by SDS-PAGE/CBB staining. |
Quantity | 5 μg |
Storage Requirements | Stored at -80°C. Aliquot to avoid repeated freezing and thawing. |
Gene Alias | BUB1A, BUB1L, hBUB1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction