Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bub1 Monoclonal Antibody (14H5), Active Motif™

Mouse Monoclonal Antibody
$132.55 - $397.65
Specifications
Antigen | Bub1 |
---|---|
Clone | 14H5 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-2809
![]() ![]() |
Active Motif
39736 |
10 μg |
Each for $132.55
|
|
|||||
50-199-2812
![]() ![]() |
Active Motif
39735 |
100 μg |
Each for $397.65
|
|
|||||
Description
BUB1A is a kinase involved in spindle checkpoint function. The kinase functions in part by phosphorylating a member of the miotic checkpoint complex and activating the spindle checkpoint.Specifications
Bub1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8 | |
AL022991; BUB1; BUB1 budding uninhibited by benzimidazoles 1 homolog; BUB1 mitotic checkpoint serine/threonine kinase; BUB1, mitotic checkpoint serine/threonine kinase; BUB1A; BUB1L; budding uninhibited by benzimidazoles 1 homolog; budding uninhibited by benzimidazoles 1 homolog (S. cerevisiae); C80208; D2Xrf87; hBUB1; mBUB1; Mitotic checkpoint serine/threonine-protein kinase BUB1; mitotic spindle checkpoint kinase; putative serine/threonine-protein kinase | |
BUB1 | |
IgG | |
Protein G | |
Antibody |
14H5 | |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Human, Mouse | |
O08901, O43683 | |
12235, 699 | |
This Bub1 antibody was raised against a recombinant protein corresponding to amino acids 281-419 of human Bub1. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
BUB1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title