Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bub1 Recombinant Rabbit Monoclonal Antibody (9C11)
GREENER_CHOICE

Catalog No. PIMA538345
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538345 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538345 Supplier Invitrogen™ Supplier No. MA538345
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This antibody has been tested in direct-ELISA.

The BUB1 gene encodes a serine/threonine-protein kinase that is integral to the spindle assembly checkpoint, a crucial mechanism ensuring proper chromosome alignment and segregation during mitosis. BUB1 is involved in detecting and correcting errors in kinetochore-microtubule attachments, which is vital for maintaining genomic stability. It acts by delaying the onset of anaphase until all chromosomes are correctly attached to the spindle apparatus. BUB1 is recruited to kinetochores in early mitosis, where it phosphorylates several substrates to activate other checkpoint proteins, such as BUBR1 and MAD2, thereby preventing premature progression through mitosis. Mutations or aberrations in the expression levels of BUB1 are often associated with aneuploidy and chromosomal instability, contributing to cancer progression. BUB1 is thus considered a potential target for cancer therapeutics, aiming to exploit its role in the mitotic checkpoint to selectively eliminate tumor cells through checkpoint bypass or mitotic arrest.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bub1
Applications Western Blot
Classification Recombinant Monoclonal
Clone 9C11
Concentration 1.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 150mM NaCl and 0.02% sodium azide; pH 7.4
Gene BUB1
Gene Accession No. O43683
Gene Alias Aamp; Aamp-rs; AL022991; angio associated migratory cell protein; angio-associated migratory cell protein; angio-associated migratory protein; angio-associated, migratory cell protein; AU040907; BUB1; BUB1 budding uninhibited by benzimidazoles 1 homolog; BUB1 mitotic checkpoint serine/threonine kinase; BUB1, mitotic checkpoint serine/threonine kinase; BUB1A; BUB1L; budding uninhibited by benzimidazoles 1 homolog; budding uninhibited by benzimidazoles 1 homolog (S. cerevisiae); C80208; D2Xrf87; hBUB1; mBUB1; mitotic checkpoint serine/threonine-protein kinase BUB1; mitotic spindle checkpoint kinase; putative serine/threonine-protein kinase
Gene Symbols BUB1
Host Species Rabbit
Immunogen A synthesized peptide derived from human Bub1 (aa 1-182).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 699
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.