Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bub1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30343220UL
Description
Bub1 Polyclonal specifically detects Bub1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
Bub1 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, KnockDown | |
Unconjugated | |
BUB1 budding uninhibited by benzimidazoles 1 homolog, BUB1Amitotic checkpoint serine/threonine-protein kinase BUB1, BUB1L, budding uninhibited by benzimidazoles 1 (yeast homolog), budding uninhibited by benzimidazoles 1 homolog (yeast), EC 2.7.11.1, hBUB1mitotic spindle checkpoint kinase, putative serine/threonine-protein kinase | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human Bub1 (NP_004327.1). | |
20 μL | |
Breast Cancer, Cell Cycle and Replication, Core ESC Like Genes, Mitotic Regulators, Protein Kinase, Stem Cell Markers | |
699 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction