Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bub3 Polyclonal Antibody

Bub3 Polyclonal Antibody
Supplier: Thermo Scientific PA572174
Description
Bub3 Polyclonal Antibody for Western Blot

Specifications
Bub3 | |
Polyclonal | |
Unconjugated | |
BUB3 | |
Aa2-050; AU019800; AU021329; AU043350; AW146323; bub3; BUB3 budding uninhibited by benzimidazoles 3 homolog; BUB3 mitotic checkpoint protein; BUB3, mitotic checkpoint protein; BUB3L; Bub3-PA; Bub3-PB; budding uninhibited by benomyl; budding uninhibited by benzimidazoles 3 homolog; budding uninhibited by benzimidazoles 3 homolog (S. cerevisiae); C78067; CG7581; CG7581-PA; CG7581-PB; dBUB3; Dmbub3; Dmel\CG7581; Dmel_CG7581; hBUB3; mitotic checkpoint component; Mitotic checkpoint protein BUB3; OTTHUMP00000020682; testicular tissue protein Li 27; WD repeat type I transmembrane protein A72.5 | |
Rabbit | |
Selective Precipitation | |
RUO | |
43490 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
0 | |
BUB3 | |
KLH conjugated synthetic peptide between 201-230 amino acids from Drosophila melanogaster (Fruit fly) BUB3. | |
400 μL | |
Primary | |
Drosophila | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction